missing translation for 'onlineSavingsMsg'
Learn More

cis-1,3-Cyclohexanediol 98.0+%, TCI America™

Catalog No. C23221G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23221G 1 g
C23225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23221G Supplier TCI America Supplier No. C23221G
Only null left

Chemical Identifiers

CAS 823-18-7
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key RLMGYIOTPQVQJR-OLQVQODUSA-N
Synonym cis-1,3-Dihydroxycyclohexane
PubChem CID 6432741
IUPAC Name (1S,3R)-cyclohexane-1,3-diol
SMILES C1CC(CC(C1)O)O

Specifications

Melting Point 83°C
Color White-Yellow
Boiling Point 125°C
Quantity 1 g
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,3-Cyclohexanediol
EINECSNumber (3)-2310
TSCA No