missing translation for 'onlineSavingsMsg'
Learn More

cis-1,3-Cyclohexanediamine 98.0+%, TCI America™

Catalog No. p-7125655
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28011G 1 g
C28015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28011G Supplier TCI America Supplier No. C28011G

Chemical Identifiers

CAS 26772-34-9
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
InChI Key GEQHKFFSPGPGLN-OLQVQODUSA-N
Synonym cis-1,3-Diaminocyclohexane
PubChem CID 6998900
IUPAC Name (1R,3S)-cyclohexane-1,3-diamine
SMILES C1CC(CC(C1)N)N

Specifications

Color Colorless
Boiling Point 198°C
Quantity 1 g
UN Number 2735
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material cis-1,3-Cyclohexanediamine
EINECSNumber (3)-3871
TSCA No