missing translation for 'onlineSavingsMsg'
Learn More

cis-1,3-Bis(aminomethyl)cyclohexane 98.0+%, TCI America™

Catalog No. p-7123393
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41761G 1 g
B41765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41761G Supplier TCI America Supplier No. B41761G
Only null left

Chemical Identifiers

CAS 10340-00-8
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
InChI Key QLBRROYTTDFLDX-OCAPTIKFSA-N
Synonym cis-1,3-Di(aminomethyl)cyclohexane, cis-Hexahydro-m-xylylenediamine
PubChem CID 6951508
IUPAC Name [(1S,3R)-3-(aminomethyl)cyclohexyl]methanamine
SMILES C1CC(CC(C1)CN)CN

Specifications

Color Colorless
Boiling Point 72°C
Quantity 1 g
UN Number 2735
Formula Weight 142.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material cis-1,3-Bis(aminomethyl)cyclohexane
EINECSNumber (3)-2279
TSCA No