missing translation for 'onlineSavingsMsg'
Learn More

(±)-cis-1,2-Diaminocyclohexane, 97%, Thermo Scientific™

Catalog No. AC248660010
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC248660010 1g Glass bottle
AC248660050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC248660010 Supplier Thermo Scientific Chemicals Supplier No. 248660010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1436-59-5
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00063746
InChI Key SSJXIUAHEKJCMH-UHFFFAOYNA-N
Synonym cis-1,2-diaminocyclohexane, cis-1,2-cyclohexanediamine, 1r,2s-cyclohexane-1,2-diamine, unii-r0zn6k26ep, cis-cyclohexane-1,2-diamine, 1,2-diaminocyclohexane, cis, r0zn6k26ep, 1,2-cyclohexanediamine, 1r,2s-rel, 1,2-cyclohexanediamine #, cis-1,2-cyclohexandiamine
PubChem CID 342917
IUPAC Name (1S,2R)-cyclohexane-1,2-diamine
SMILES NC1CCCCC1N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 0.9520g/mL
Boiling Point 92.0°C (14.0 mbar)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4930 to 1.4960 (20°C, 589nm)
Quantity 1g
Specific Gravity 0.952
Formula Weight 114.19
Percent Purity 97%
Chemical Name or Material (±)-cis-1, 2-Diaminocyclohexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only