missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclohexanediol 98.0+%, TCI America™

Catalog No. C18021G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C18021G Supplier TCI America Supplier No. C18021G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1792-81-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00064944
InChI Key PFURGBBHAOXLIO-OLQVQODUSA-N
Synonym cis-1,2-cyclohexanediol, cis-cyclohexane-1,2-diol, grandidentol, cis-1,2-dihydroxycyclohexane, 1r,2s-cyclohexane-1,2-diol, 1,2-cyclohexanediol, cis, 1,2-cyclohexanediol, cis-8ci 9ci, 1,2-cyclohexanediol, 1r,2s-rel, cis-1,2-cyclohexandiol, 1alpha,2alpha-cyclohexanediol
PubChem CID 92903
ChEBI CHEBI:32329
IUPAC Name (1R,2S)-cyclohexane-1,2-diol
SMILES C1CCC(C(C1)O)O

Specifications

Melting Point 100°C
Color White
Boiling Point 116°C
Quantity 1 g
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,2-Cyclohexanediol
EINECSNumber (3)-2310
TSCA No