missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclohexanediol, 99%

Catalog No. AC300970050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC300970050 5 g Glass bottle
AC300970010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC300970050 Supplier Thermo Scientific Chemicals Supplier No. 300970050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1792-81-0 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1792-81-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00064944
InChI Key PFURGBBHAOXLIO-OLQVQODUSA-N
Synonym cis-1,2-cyclohexanediol, cis-cyclohexane-1,2-diol, grandidentol, cis-1,2-dihydroxycyclohexane, 1r,2s-cyclohexane-1,2-diol, 1,2-cyclohexanediol, cis, 1,2-cyclohexanediol, cis-8ci 9ci, 1,2-cyclohexanediol, 1r,2s-rel, cis-1,2-cyclohexandiol, 1alpha,2alpha-cyclohexanediol
PubChem CID 92903
ChEBI CHEBI:32329
IUPAC Name (1R,2S)-cyclohexane-1,2-diol
SMILES C1CCC(C(C1)O)O

Specifications

Melting Point 98°C to 101°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(OH)2
Quantity 5 g
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 116.16
Percent Purity 99%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material cis-1, 2-Cyclohexanediol
Show More Show Less

RUO – Research Use Only