missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclohexanedicarboxylic Anhydride 99.0+%, TCI America™

Catalog No. C047825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C047825G 25 g
C0478100G 100 g
C0478500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C047825G Supplier TCI America Supplier No. C047825G

Chemical Identifiers

CAS 13149-00-3
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
MDL Number MFCD00005926
InChI Key MUTGBJKUEZFXGO-OLQVQODUSA-N
Synonym cis-hexahydroisobenzofuran-1,3-dione, cis-1,2-cyclohexanedicarboxylic anhydride, cis-hexahydrophthalic anhydride, unii-b52z7tnv87, cis-cyclohexane-1,2-dicarboxylic anhydride, hexahydrophthalic anhydride, cis, 3ar,7as-hexahydro-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, 3ar,7as-rel, 3ar,7as-hexahydroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, hexahydro-, cis
PubChem CID 641660
IUPAC Name (3aS,7aR)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES C1CCC2C(C1)C(=O)OC2=O

Specifications

Color White
Boiling Point 160°C
Quantity 25 g
Formula Weight 154.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material cis-1,2-Cyclohexanedicarboxylic Anhydride
EINECSNumber (3)-2416
TSCA No