missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec™ Circular Specimen Chucks for SHANDON Cryostats
Description
- Round, aluminum specimen chucks
Specifications
Specifications
| For Use With (Equipment) | SHANDON Cryostats |
| Length (Metric) | 40 mm |
| Color | Purple |