missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Circular Specimen Chucks for SHANDON Cryostats

Catalog No. 22445473
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Length (Metric):
30 mm
40 mm
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (Metric)
22445473 Pink 30 mm
22445471 Blue 30 mm
22445558 Green 30 mm
22445361 Orange 30 mm
22445622 Purple 30 mm
22445607 Red 30 mm
22445671 White 30 mm
22445770 Yellow 30 mm
22445258 Blue 40 mm
22445682 Green 40 mm
22445769 Orange 40 mm
22445606 Pink 40 mm
22445803 Purple 40 mm
22445835 Red 40 mm
22445334 White 40 mm
22445670 Yellow 40 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 22445473 Supplier Mopec™ Supplier No. SP143PK
Only null left

Available in multiple sizes and colors for use with SHANDON Cryostats.

  • Round, aluminum specimen chucks

Specifications

For Use With (Equipment) SHANDON Cryostats
Length (Metric) 30 mm
Color Pink