missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats

Catalog No. 22445748
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Length (Metric):
22 mm
25 mm
30 mm
40 mm
55 mm
40 product options available for selection
Product selection table with 40 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (Metric)
22445748 Blue 40 mm
22445492 Blue 22 mm
22445797 Green 22 mm
22445321 Orange 22 mm
22445796 Pink 22 mm
22445487 Purple 22 mm
22445320 Red 22 mm
22445652 White 22 mm
22445330 Yellow 22 mm
22445331 Blue 25 mm
22445613 Green 25 mm
22445604 Orange 25 mm
22445282 Pink 25 mm
22445281 Purple 25 mm
22445788 Red 25 mm
22445684 White 25 mm
22445605 Yellow 25 mm
22445776 Blue 30 mm
22445786 Green 30 mm
22445609 Orange 30 mm
22445283 Pink 30 mm
22445777 Purple 30 mm
22445341 Red 30 mm
22445738 White 30 mm
22445712 Yellow 30 mm
22445880 Green 40 mm
22445877 Orange 40 mm
22445581 Pink 40 mm
22445879 Purple 40 mm
22445806 Red 40 mm
22445545 White 40 mm
22445559 Yellow 40 mm
22445352 Blue 55 mm
22445397 Green 55 mm
22445687 Orange 55 mm
22445490 Pink 55 mm
22445732 Purple 55 mm
22445254 Red 55 mm
22445731 White 55 mm
22445511 Yellow 55 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 40 options available.
40 options
Catalog No. 22445748 Supplier Mopec™ Supplier No. SP104BL
Only null left

Available in multiple sizes and colors for use with Leica, TBS, and Tanner Cryostats.

  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats

Specifications

For Use With (Equipment) Leica, TBS, and Tanner Cryostats
Length (Metric) 40 mm
Color Blue