Learn More
Mopec™ Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats
Available in multiple sizes and colors for use with Leica, TBS, and Tanner Cryostats.
Supplier:  Mopec™ SP105YE
 View more versions of this product
Catalog No. 22445511
Only null left
Description
Description
  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats
Specifications
Chemical Identifiers
Leica, TBS, and Tanner Cryostats
Specifications
Leica, TBS, and Tanner Cryostats
55 mm
Yellow
Product Suggestions
Product Suggestions