Learn More
Fisherbrand™ Circular Cover Glasses
Made of the finest optical borosilicate glass, with uniform thickness and size.
Supplier: Fisherbrand™ 18CIR2602811T
Description
All Circular Cover Slips feature a high-spectral transmission quality and finely adaptive refractive index suited for microscopy. Sold by weight, comes in 1 oz. packs.
- Manufactured with colorless borosilicate glass
- Available in a variety of sizes and thicknesses
Chemical Identifiers
| Circle | |
| Borosilicate Glass | |
| Microscopy, Histology, Cytology, Hematology, Microbiology | |
| 1 oz./Pk. |
Specifications
| Circle | |
| 0.19 to 0.23 mm (#2) | |
| Microscopy, Histology, Cytology, Hematology, Microbiology |
| Borosilicate Glass | |
| 18 mm | |
| 1 oz./Pk. |