Learn More
Fisherbrand™ Circular Cover Glasses
Made of the finest optical borosilicate glass, with uniform thickness and size.
Supplier:  Fisherbrand™ 15CIR1602811T
 View more versions of this product
Catalog No. 12541003CA
Only null left
Description
Description
All Circular Cover Slips feature a high-spectral transmission quality and finely adaptive refractive index suited for microscopy. Sold by weight, comes in 1 oz. packs.
  • Manufactured with colorless borosilicate glass
  • Available in a variety of sizes and thicknesses
Specifications
Chemical Identifiers
Circle
Borosilicate Glass
Microscopy, Histology, Cytology, Hematology, Microbiology
1 oz./Pk.
Specifications
Circle
0.13 to 0.17 mm (#1)
Microscopy, Histology, Cytology, Hematology, Microbiology
Borosilicate Glass
15 mm
1 oz./Pk.
Product Suggestions
Product Suggestions