missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin 98.0+%, TCI America™

Catalog No. p-7125498
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C25105G 5 g
C251025G 25 g
2 options

Catalog No. C25105G

Supplier: TCI America C25105G

Only null left

Chemical Identifiers

CAS 85721-33-1
Molecular Formula C17H18FN3O3
Molecular Weight (g/mol) 331.347
MDL Number MFCD00185755
InChI Key MYSWGUAQZAJSOK-UHFFFAOYSA-N
Synonym ciprofloxacin, ciprofloxacine, ciprobay, ciproxan, ciprofloxacina, cipro, ciprofloxacino, ciprofloxacinum, cipro iv, bacquinor
PubChem CID 2764
ChEBI CHEBI:100241
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)N4CCNCC4)F)C(=O)O

Specifications

Melting Point 265°C
Color White
Quantity 5 g
Formula Weight 331.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ciprofloxacin
RTECSNumber VB1993800
TSCA No