missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015692
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015692 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015692 Supplier Merck Emd Millipore Supplier No. PHR11671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Ciprofloxacin is grouped under the class of second generation of quinolone analogs of nalidixic acid that exhibits high potency, minimal toxicity and a broad-spectrum antibacterial activity. It can be administered both parenterally and orally. It finds applications in the treatment of broad range of infections of the urinary tract, respiratory tract and gastrointestinal tract, as well as skin and soft tissue infections. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 85721-33-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C17H18 FN3O3
MDL Number MFCD00185755
Synonym 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid; Ciprobay
Type Certified Reference Material
Quantity 1 g
Formula Weight 331.34
Grade Certified Reference Material