missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin, 98%

Catalog No. AAJ6131706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6131706 5 g
AAJ6131714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6131706 Supplier Thermo Scientific Chemicals Supplier No. J6131706

An anti-bacterial agent effective against anaerobic bacteria | CAS: 85721-33-1 | C17H18FN3O3 | 331.347 g/mol

Ciprofloxacin, inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ciprofloxacin, inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome.

Solubility
Soluble in 0.1N HCl at 25mg/ml. Poorly soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 85721-33-1
Molecular Formula C17H18FN3O3
Molecular Weight (g/mol) 331.347
MDL Number MFCD00185755
InChI Key MYSWGUAQZAJSOK-UHFFFAOYSA-N
PubChem CID 2764
ChEBI CHEBI:100241
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)N4CCNCC4)F)C(=O)O

Specifications

Type Ciprofloxacin
Physical Form Powder
Melting Point 253°C to 257°C
Color White
Quantity 5 g
Beilstein 3568352
Merck Index 14,2314
Solubility Information Soluble in 0;1N HCl at 25mg/ml; Poorly soluble in DMSO
Formula Weight 331.3
Percent Purity 98%
Chemical Name or Material Ciprofloxacin
Show More Show Less
Precautionary Statement P260-P271-P304+P340-P312-P314-P501c
RTECSNumber VB1993800
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only