missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ CIP Additive Detergent Booster and Defoamer

Catalog No. 17355275
Change view
Click to view available options
Size:
55 gal.
300 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17355275 55 gal.
17355273 300 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17355275 Supplier STERIS™ Supplier No. 1D0301
Only null left

CIP Additive Detergent Booster and Defoamer is a neutral pH detergent additive for use in boosting the cleaning ability of either alkaline or acidic cleaners. CIP Additive Detergent Booster and Defoamer can be used to supplement surfactants and to enhance cleaning. pH - undiluted 8.57, typical.

CIP Additive Detergent Booster and Defoamer is specially formulated to enhance the cleaning performance of STERIS formulated detergents in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

HOW CIP ADDITIVE WORKS

CIP Additive Detergent is a proprietary blend of surfactants and other performance-enhancing ingredients. This low foaming product removes a wide range of process residues and is ideal for use in CIP, COP, and manual applications.

WHY CIP ADDITIVE?

  • Productivity - Enhances the performance of other formulated detergents with additional sufactants.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics when used in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Safety - Enhanced operator safety due to moderate pH.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.

Specifications

Product Type Detergent Additive
Color-coded No
Form Clear Liquid
pH 8.57 (Undiluted)
Scent Unscented
Sterility Non-sterile
Size 55 gal.