missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ CIP 150 Alkaline Process and Research Cleaner

CIP 150 Alkaline Process and Research Cleaner is a specially formulated spray-wash compound that combines an excellent alkaline detergent system with the protein cleaning properties of chlorine. It utilizes improved chemistry, significantly extending the stability of active chlorine. pH - (1%) 12.2.

$4,653.00 - $40,058.23

Specifications

Product Type Alkaline Cleaner
Color-coded No
Form Clear Liquid
pH 12.2 (1%)
Scent Unscented
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17355269
View Documents Promotion Details
STERIS™
1D1502
30 gal.
Each for $4,653.00
Only null left
 
17355267
View Documents Promotion Details
STERIS™
1D15D3
300 gal. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

CIP 150 Alkaline Process and Research Cleaner is specially formulated to meet the unique cleaning demands found in biopharmaceutical manufacturing. The blend of alkalinity, surfactants, and oxidizing agents were developed for the most challenging protein-based process residues.

HOW CIP 150 ALKALINE DETERGENT WORKS

CIP 150 Alkaline Detergent is a proprietary blend of potassium hydroxide, sodium hypochlorite (oxidizing agent), surfactants and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in CIP, COP and manual applications.

WHY CIP 150 ALKALINE DETERGENT?

  • Flexibility - Effective cleaning against a wide variety of organic and proteinaceous process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
Specifications

Specifications

Alkaline Cleaner
Clear Liquid
Unscented
No
12.2 (1%)
Non-sterile
Videos
SDS
Documents

Documents