missing translation for 'onlineSavingsMsg'
Learn More

STERIS CIP 100™ Alkaline Process and Research Cleaner

Catalog No. 361007319
Change view
Click to view available options
Volume (English):
1 gal.
55 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
361007319 1 gal.
296364286 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361007319 Supplier STERIS Supplier No. 1D1008

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specially formulated to meet the unique cleaning demands found in industries that require an effective, validatable detergent.

  • A proprietary blend of potassium hydroxide, an advanced surfactant system, and other performance-enhancing ingredients that provide multiple cleaning mechanisms
  • Removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants
  • Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Ideal for use in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • For use in pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries that require an effective, validatable detergent
  • Comprehensive support documentation package includes specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Manufactured in a cGMP compliant, FDA registered, and ISO 9001 certified facility

Specifications

Product Type Detergent
Composition A Proprietary Blend of Potassium Hydroxide (KOH)
Container Type Bottle
For Use With (Application) Clean-in-Place (CIP), Clean-Out-of-Place (COP), and Manual Applications
Quantity 4/Cs.
Volume (English) 1 gal.