missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl alcohol, 98%

Catalog No. p-7020912
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1302530 250 g
AAA130250B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1302530 Supplier Thermo Scientific Chemicals Supplier No. A1302530
Only null left

CAS: 104-54-1 | C9H10O | 134.178 g/mol

Cinnamyl alcohol is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in perfumery and as well as a deodorant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamyl alcohol is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in perfumery and as well as a deodorant.

Solubility
Soluble in water 1.8 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 104-54-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00002921
InChI Key OOCCDEMITAIZTP-QPJJXVBHSA-N
Synonym cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol
PubChem CID 5315892
ChEBI CHEBI:33227
IUPAC Name (E)-3-phenylprop-2-en-1-ol
SMILES C1=CC=C(C=C1)C=CCO

Specifications

Melting Point 32°C to 35°C
Density 1.044
Boiling Point 257°C to 258°C
Flash Point 126°C (258°F)
Odor Pleasant
Linear Formula C6H5CH=CHCH2OH
Quantity 250 g
Beilstein 1903999
Merck Index 14,2302
Solubility Information Soluble in water 1.8g/L (20°C).
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material Cinnamyl alcohol
Show More Show Less
Hazard Category H302-H317-H335
Hazard Statement GHS H Statement
H302-H315-H319-H317
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c
EINECSNumber 203-212-3
RTECSNumber GE2200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only