missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cinnamyl-3,4-dihydroxy-alpha-cyanocinnamate, 98%

CAS: 132465-11-3 | C19H15NO4, C19H15NO4 | 321.33 g/mol

Supplier:  Thermo Scientific Chemicals J67167MA

Catalog No. AAJ67167MA


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

10 mg
98%
C19H15NO4
CDC;
Powder
OC1=C(O)C=C(C=C(C#N)C(=O)OCC=CC2=CC=CC=C2)C=C1
321.33
Yellow-orange
132465-11-3
321.33
Cinnamyl-3,4-dihydroxy-alpha-cyanocinnamate, 98%
XGHYFEJMJXGPGN-UHFFFAOYSA-N
3-phenylprop-2-en-1-yl 2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoate

Chemical Identifiers

132465-11-3
321.33
CDC;
OC1=C(O)C=C(C=C(C#N)C(=O)OCC=CC2=CC=CC=C2)C=C1
C19H15NO4
XGHYFEJMJXGPGN-UHFFFAOYSA-N
3-phenylprop-2-en-1-yl 2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only