Learn More
Cincinnati Surgical Lance Sterile Carbon Blades
Made from British steel, offering unique shape, size and material composition
$133.21 - $144.57
Specifications
| Blade Material | British Cold-rolled Surgical Carbon Steel |
|---|---|
| Description | Low Surface Finish |
| Sterility | Sterile |
| Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
|
19200268
|
Cincinnati Surgical
92010 |
# 10 |
Pack of 100 Each for $133.21
|
|
|||||
|
19200269
|
Cincinnati Surgical
92011 |
# 11 |
Pack of 100 Each for $136.07
|
|
|||||
|
19200270
|
Cincinnati Surgical
92012 |
# 12 |
Pack of 100 Each for $133.21
|
|
|||||
|
19200271
|
Cincinnati Surgical
92015T |
# 15 |
Pack of 100 Each for $133.21
|
|
|||||
|
19200272
|
Cincinnati Surgical
92020 |
Pack of 100 Each for $138.89
|
|
||||||
|
19200273
|
Cincinnati Surgical
92021 |
# 21 |
Pack of 100 Each for $144.57
|
|
|||||
|
19200274
|
Cincinnati Surgical
92022 |
# 22 |
Pack of 100 Each for $138.89
|
|
|||||
Description
- 100% visual inspection
- Wide grinding angle for high durability
- Low surface finish
Certifications
CE Marked
Specifications
| British Cold-rolled Surgical Carbon Steel | |
| Low Surface Finish | |
| Sterile |