Learn More
Cincinnati Surgical Swann Morton Sterile Stainless Steel Blades and Scalpels
Catalog No. 19200241
Change view
Click to view available options
Size:
# 10
# 10A
# 11
# 12
# 15
# 20
# 21
# 22
# 23
# 25A
Product Type:
Blade
Scalpel
Scalpel Blade
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Product Type
19200241 # 20 Blade
19200237 # 10 Blade
19200238 # 11 Blade
19200239 # 12 Blade
19200240 # 15 Blade
19200242 # 21 Blade
19200243 # 22 Blade
19200244 # 25A Blade
19200249 # 10A Blade
19200222 # 10 Scalpel
19200223 # 11 Scalpel Blade
19200224 # 15 Scalpel Blade
19200225 # 20 Scalpel Blade
19200226 # 21 Scalpel
19200227 # 22 Scalpel
19200228 # 23 Scalpel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 19200241 Supplier Cincinnati Surgical Supplier No. 01SM20
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use in wide variety of medical applications, including Dermaplaning
  • Blades packaged with VCI Paper to protect blade from corrosion
  • Scalpels individually packaged in transparent thermos formed tray
  • 200% visual inspection
  • Strongest/Smallest foil package available
  • Narrow grinding angle resulting in increased initial keenness
  • High surface finish
  • Cellophane overwrap
  • Extended range
Specifications
Blade Material Stainless Steel
Product Type Blade
Size # 20
Sterility Sterile
Packaging Type Packaged with VCI Paper to protect Blade from Corrosion