Learn More
Cincinnati Surgical Swann Morton Sterile Stainless Steel Blades and Scalpels
Description
For use in wide variety of medical applications, including Dermaplaning
- Blades packaged with VCI Paper to protect blade from corrosion
- Scalpels individually packaged in transparent thermos formed tray
- 200% visual inspection
- Strongest/Smallest foil package available
- Narrow grinding angle resulting in increased initial keenness
- High surface finish
- Cellophane overwrap
- Extended range
Specifications
Specifications
| Blade Material | Stainless Steel |
| Product Type | Blade |
| Size | # 10 |
| Sterility | Sterile |
| Packaging Type | Packaged with VCI Paper to protect Blade from Corrosion |