Learn More
Cincinnati Surgical Sterile-Stainless Steel Blades and Scalpels
Catalog No. 19200209
Change view
Click to view available options
Size:
# 10
# 11
# 15
# 21
# 22
Quantity:
10/Pk.
Product Type:
Safety Scalpel
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Quantity Product Type
19200209 # 22 10/Pk. Safety Scalpel
19200204 # 10 10/Pk. Safety Scalpel
19200205 # 11 10/Pk. Safety Scalpel
19200206 # 15 10/Pk. Safety Scalpel
19200208 # 21 10/Pk. Safety Scalpel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19200209 Supplier Cincinnati Surgical Supplier No. 0522
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Packaged with VCI Paper to protect the blade from corrosion.
  • Made from British stainless steel
  • 200% visual inspection
  • Narrow grinding angle resulting in increased initial keenness
  • Medium surface finish
  • Strong foil package
  • Handles are made of plastic
Specifications
Blade Material British Stainless Steel
Product Type Safety Scalpel
Size # 22
For Use With (Equipment) Flexible Blade Cover
Description Medium Surface Finish
Sterility Sterile
Quantity 10/Pk.