Learn More
Cincinnati Surgical Sterile-Stainless Steel Blades and Scalpels
Packaged with VCI Paper to protect the blade from corrosion.
Supplier: Cincinnati Surgical 0521
Description
- Made from British stainless steel
- 200% visual inspection
- Narrow grinding angle resulting in increased initial keenness
- Medium surface finish
- Strong foil package
- Handles are made of plastic
Chemical Identifiers
| British Stainless Steel | |
| Medium Surface Finish | |
| Sterile |
Specifications
| British Stainless Steel | |
| # 21 | |
| Medium Surface Finish | |
| 10/Pk. |
| Safety Scalpel | |
| Flexible Blade Cover | |
| Sterile |