Learn More
Cincinnati Surgical Lance Sterile Stainless Steel Blades and Scalpels
Made from British cold-rolled surgical stainless steel, offering unique shape, size and material composition
Supplier: Cincinnati Surgical 92115T
Description
- 100% visual inspection
- Wide grinding angle for high durability
- Low surface finish
- Flexible blade cover included with each scalpel
- Scalpels individually packaged
Certifications
CE Marked
Chemical Identifiers
| Stainless Steel | |
| Sterile |
Specifications
| Stainless Steel | |
| # 15 | |
| Sterile |
| Blade | |
| Low Surface Finish Blade | |
| 10/Pk. |