Learn More
Cincinnati Surgical Lance Sterile Stainless Steel Blades and Scalpels
Made from British cold-rolled surgical stainless steel, offering unique shape, size and material composition
Supplier:  Cincinnati Surgical 92420
 View more versions of this product
Catalog No. 19200265
Only null left
Description
Description
  • 100% visual inspection
  • Wide grinding angle for high durability
  • Low surface finish
  • Flexible blade cover included with each scalpel
  • Scalpels individually packaged
Certifications
CE Marked
Specifications
Chemical Identifiers
Stainless Steel
Sterile
Specifications
Stainless Steel
# 20
Sterile
Scalpel
Low Surface Finish, Green Handle, Flexible
10 Pk.
Product Suggestions
Product Suggestions