missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Cinchonine | |
| 261°C | |
| MFCD00064372 | |
| +-quinolin-4-yl 5-vinylquinuclidin-2-yl methanol, 5r-5-vinylquinuclidin-2-yl 1s-4-quinolylmethan-1-ol | |
| [H][C@@]1(CN2CCC1C[C@]2([H])[C@@H](O)C1=CC=NC2=CC=CC=C12)C=C | |
| 294.40 | |
| 294.40 | |
| Crystalline Powder |
| 118-10-5 | |
| C19H22N2O | |
| 200 g | |
| KMPWYEUPVWOPIM-FRYPIZGFNA-N | |
| (S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](quinolin-4-yl)methanol | |
| 21862290 | |
| ≥98.0% (T) |
Chemical Identifiers
| 118-10-5 | |
| 294.40 | |
| KMPWYEUPVWOPIM-FRYPIZGFNA-N | |
| 21862290 | |
| [H][C@@]1(CN2CCC1C[C@]2([H])[C@@H](O)C1=CC=NC2=CC=CC=C12)C=C |
| C19H22N2O | |
| MFCD00064372 | |
| +-quinolin-4-yl 5-vinylquinuclidin-2-yl methanol, 5r-5-vinylquinuclidin-2-yl 1s-4-quinolylmethan-1-ol | |
| (S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](quinolin-4-yl)methanol |
Safety and Handling
EINECSNumber : (9)-0500
RTECSNumber : GD3500000
TSCA : Yes