missing translation for 'onlineSavingsMsg'
Learn More

Cinchonine 98.0+%, TCI America™

Catalog No. C035025G
Change view
Click to view available options
Quantity:
25 g
200 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C035025G 25 g
C0350200G 200 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C035025G Supplier TCI America Supplier No. C035025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118-10-5
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00064372
InChI Key KMPWYEUPVWOPIM-FRYPIZGFNA-N
Synonym +-quinolin-4-yl 5-vinylquinuclidin-2-yl methanol, 5r-5-vinylquinuclidin-2-yl 1s-4-quinolylmethan-1-ol
PubChem CID 21862290
IUPAC Name (S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](quinolin-4-yl)methanol
SMILES [H][C@@]1(CN2CCC1C[C@]2([H])[C@@H](O)C1=CC=NC2=CC=CC=C12)C=C

Specifications

Melting Point 261°C
Quantity 25 g
Formula Weight 294.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cinchonine
EINECSNumber (9)-0500
RTECSNumber GD3500000
TSCA Yes