missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cinchonine, 98+%

Catalog No. AC227815000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227815000 500 g Glass bottle
AC227810250 25 g Glass bottle
AC227811000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227815000 Supplier Thermo Scientific Chemicals Supplier No. 227815000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 118-10-5 | C19H22N2O | 294.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-10-5
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00064372
InChI Key KMPWYEUPVWOPIM-FRYPIZGFNA-N
Synonym +-quinolin-4-yl 5-vinylquinuclidin-2-yl methanol, 5r-5-vinylquinuclidin-2-yl 1s-4-quinolylmethan-1-ol
PubChem CID 21862290
IUPAC Name [(5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol
SMILES [H][C@@]1(CN2CCC1C[C@]2([H])[C@@H](O)C1=CC=NC2=CC=CC=C12)C=C

Specifications

Melting Point 255.0°C to 265.0°C
Color White to Beige
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 500 g
Beilstein 23, V,12, 406
Fieser 06,501; 13,264
Merck Index 15,229
Specific Rotation Condition +251.00 (20.00°C c=2,0.2M HCl)
Specific Rotation + 251.00
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/60mL alcohol - 1g/25mL boiling alcohol, 1g/110mL chloroform - 1g/500mL ether
Formula Weight 294.4
Percent Purity 99%
Physical Form Micro-crystalline Powder
Chemical Name or Material Cinchonine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If s
EINECSNumber 204-234-6

RUO – Research Use Only