missing translation for 'onlineSavingsMsg'
Learn More

(+)-Cinchonine, 98+%, cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine

Catalog No. AAA1752314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1752314 25 g
AAA1752322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1752314 Supplier Thermo Scientific Chemicals Supplier No. A1752314

CAS: 118-10-5 | C19H22N2O | 294.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-10-5
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00064372
InChI Key KMPWYEUPVWOPIM-FRYPIZGFNA-N
Synonym +-quinolin-4-yl 5-vinylquinuclidin-2-yl methanol, 5r-5-vinylquinuclidin-2-yl 1s-4-quinolylmethan-1-ol
PubChem CID 21862290
IUPAC Name [(5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol
SMILES [H][C@@]1(CN2CCC1C[C@]2([H])[C@@H](O)C1=CC=NC2=CC=CC=C12)C=C

Specifications

Melting Point 253°C to 258°C
Quantity 25 g
Beilstein 89689
Sensitivity Light Sensitive
Merck Index 14,2287
Optical Rotation +225° (c=0.5 in Ethanol)
Formula Weight 294.39
Percent Purity ≥98%
Assay cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine
Chemical Name or Material (+)-Cinchonine
Hazard Category H302-H317
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
EINECSNumber 204-234-6
RTECSNumber GD3500000
TSCA Yes

RUO – Research Use Only