missing translation for 'onlineSavingsMsg'
Learn More

Cinchonidine 98.0+%, TCI America™

Catalog No. p-7124363
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
C034725G 25 g
C0347100G 100 g
2 options

Catalog No. C034725G

Supplier: TCI America C034725G

Only null left

Chemical Identifiers

CAS 485-71-2
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00006783
InChI Key KMPWYEUPVWOPIM-UHFFFAOYNA-N
Synonym cinchonidine
PubChem CID 45358337
IUPAC Name {5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(quinolin-4-yl)methanol
SMILES OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 25 g
Formula Weight 294.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cinchonidine
EINECSNumber (9)-0501
RTECSNumber GD2975000
TSCA Yes