missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Cinchonidine, 99% (total base), may cont. up to 5% quinine
Description
It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.
Solubility
Insoluble in water. Soluble in alcohol and chloroform.
Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.
Solubility
Insoluble in water. Soluble in alcohol and chloroform.
Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 201°C to 206°C |
| Quantity | 25 g |
| Beilstein | 89690 |
| Sensitivity | Light sensitive |
| Merck Index | 14,2286 |
| Solubility Information | Insoluble in water. Soluble in alcohol and chloroform. |
| Optical Rotation | −109° (c=0.5 in Ethanol) |
| IUPAC Name | {5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(quinolin-4-yl)methanol |
| Formula Weight | 294.39 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H302-H317 |
| Hazard Statement | GHS H Statement H334-H302-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c |
| EINECSNumber | 207-622-3 |
| RTECSNumber | GD3500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only