missing translation for 'onlineSavingsMsg'
Learn More

(-)-Cinchonidine, 99% (total base), may cont. up to 5% quinine

Catalog No. AAA1879614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1879614 25 g
AAA1879622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1879614 Supplier Thermo Scientific Chemicals Supplier No. A1879614

CAS: 485-71-2 | C19H22N2O | 294.40 g/mol

It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.

Solubility
Insoluble in water. Soluble in alcohol and chloroform.

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 485-71-2
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00006783
InChI Key KMPWYEUPVWOPIM-UHFFFAOYNA-N
Synonym cinchonidine
PubChem CID 45358337
SMILES OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 201°C to 206°C
Quantity 25 g
Beilstein 89690
Sensitivity Light sensitive
Merck Index 14,2286
Solubility Information Insoluble in water. Soluble in alcohol and chloroform.
Optical Rotation −109° (c=0.5 in Ethanol)
IUPAC Name {5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(quinolin-4-yl)methanol
Formula Weight 294.39
Percent Purity 99%
Assay (total base), may cont. up to 5% quinine
Chemical Name or Material (-)-Cinchonidine
Show More Show Less
Hazard Category H302-H317
Hazard Statement GHS H Statement
H334-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
EINECSNumber 207-622-3
RTECSNumber GD3500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only