missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cinchonidine, 98.5-101%

Catalog No. AC110330250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC110330250 25 g
AC110331000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110330250 Supplier Thermo Scientific Chemicals Supplier No. 110330250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Cinchonidine, CAS # 485-71-2, is a naturally occurring alkaloid that may be used in asymmetric synthesis and exhibits anti-malarial activity | CAS: 485-71-2 | C19H22N2O | 294.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cinchonidine is an alkaloid naturally occurring in the bark of Cinchona strubs that has activity against Plasmodium spp
  • Cinchonidine is a diastereoisomer of cinchonine and can be used to promote chirality in asymmetric or enantioselective synthesis of organic compounds

Applications

  • Cinchonidine has been used as a starting material in the synthesis of anti-malarial cinchona alkaloid conjugates
  • Has been shown to potently induce γ-globin mRNA and fetal hemoglobin in erythroid progenitor cells and may be useful in preclinical studies of β-thalassemia

Chemical Identifiers

CAS 485-71-2
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00006783
InChI Key KMPWYEUPVWOPIM-UHFFFAOYNA-N
Synonym cinchonidine
PubChem CID 45358337
IUPAC Name [(2R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol
SMILES OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 200.0°C to 203.0°C
Color White to Cream
Loss on Drying 1% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98.5-101%
Packaging Glass bottle
Quantity 25 g
Beilstein 23, 420
Merck Index 15, 2289
Specific Rotation − 115.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and chloroform
Formula Weight 294.39
Percent Purity 98.5-101%
Physical Form Crystalline Powder
Chemical Name or Material Cinchonidine, 98.5-101%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 207-622-3
RTECSNumber GD2975000
TSCA TSCA

RUO – Research Use Only