missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cilnidipine 98.0+%, TCI America™
$160.30 - $477.19
Chemical Identifiers
| CAS | 132203-70-4 |
|---|---|
| Molecular Formula | C27H28N2O7 |
| Molecular Weight (g/mol) | 492.528 |
| MDL Number | MFCD00865853 |
| InChI Key | KJEBULYHNRNJTE-DHZHZOJOSA-N |
| Synonym | cilnidipine, cinalong, atelec, siscard, cilnidipine inn, cinaldipine, 3-cinnamyl 5-2-methoxyethyl 2,6-dimethyl-4-3-nitrophenyl-1,4-dihydropyridine-3,5-dicarboxylate, atelec tn, +--e-cinnamyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate, 1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-3,5-pyridinedicarboxylic acid 2-methoxyethyl 2e-3-phenyl-2-propenyl ester |
| PubChem CID | 5282138 |
| ChEBI | CHEBI:31399 |
| IUPAC Name | 3-O-(2-methoxyethyl) 5-O-[(E)-3-phenylprop-2-enyl] 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| SMILES | CC1=C(C(C(=C(N1)C)C(=O)OCC=CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OCCOC |
Chemical Identifiers
| 132203-70-4 | |
| 492.528 | |
| KJEBULYHNRNJTE-DHZHZOJOSA-N | |
| 5282138 | |
| 3-O-(2-methoxyethyl) 5-O-[(E)-3-phenylprop-2-enyl] 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| C27H28N2O7 | |
| MFCD00865853 | |
| cilnidipine, cinalong, atelec, siscard, cilnidipine inn, cinaldipine, 3-cinnamyl 5-2-methoxyethyl 2,6-dimethyl-4-3-nitrophenyl-1,4-dihydropyridine-3,5-dicarboxylate, atelec tn, +--e-cinnamyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate, 1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-3,5-pyridinedicarboxylic acid 2-methoxyethyl 2e-3-phenyl-2-propenyl ester | |
| CHEBI:31399 | |
| CC1=C(C(C(=C(N1)C)C(=O)OCC=CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OCCOC |
Specifications
| 132203-70-4 | |
| Yellow | |
| MFCD00865853 | |
| cilnidipine, cinalong, atelec, siscard, cilnidipine inn, cinaldipine, 3-cinnamyl 5-2-methoxyethyl 2,6-dimethyl-4-3-nitrophenyl-1,4-dihydropyridine-3,5-dicarboxylate, atelec tn, +--e-cinnamyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate, 1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-3,5-pyridinedicarboxylic acid 2-methoxyethyl 2e-3-phenyl-2-propenyl ester | |
| CC1=C(C(C(=C(N1)C)C(=O)OCC=CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OCCOC | |
| 492.528 | |
| CHEBI:31399 | |
| ≥98.0% (HPLC) | |
| Cilnidipine |
| 110°C | |
| C27H28N2O7 | |
| 1 g | |
| KJEBULYHNRNJTE-DHZHZOJOSA-N | |
| 3-O-(2-methoxyethyl) 5-O-[(E)-3-phenylprop-2-enyl] 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate | |
| 5282138 | |
| 492.53 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : US7975550
TSCA : No