missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cilnidipine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 110°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 492.53 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cilnidipine |
Safety and Handling
| RTECSNumber | US7975550 |
| TSCA | No |