missing translation for 'onlineSavingsMsg'
Learn More

α-Chymotrypsin, MP Biomedicals™

Catalog No. MP215227210
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215227210 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215227210 Supplier MP Biomedicals Inc Supplier No. 0215227210

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Preferentially catalyzes peptide bonds involving L-isomers of tyrosine, phenylalanine, and tryptophan. α-Chymotrypsin, MP Biomedicals is a serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains on the carboxyl e

α-Chymotrypsin readily acts upon amides and esters of susceptible amino acids. In addition to bonds involving aromatic amino acids, it catalyzes at a high rate the hydrolysis of bonds of leucyl, methionyl, asparaginyl, and glutamyl residues.

Chemical Identifiers

CAS 9005-12-3
Molecular Formula C7H8OSi
Molecular Weight (g/mol) 136.23
InChI Key IXKNJYSAWNJQQY-UHFFFAOYSA-N
Synonym methylphenylsilanone, polyphenylmethylsiloxane, methylphenyl oxo silane, silane, methyloxophenyl, methyl phenyl silanone
PubChem CID 3032548
IUPAC Name methyl-oxo-phenylsilane
SMILES C[Si](=O)C1=CC=CC=C1

Specifications

Quantity 10 g
Formula Weight 25 kDa (Lit.)
Physical Form Powder
Recommended Storage Store at 4°C.