missing translation for 'onlineSavingsMsg'
Learn More

α-Chymotrypsin ≥40u/mg protein, MP Biochemicals™

Catalog No. p-4685752
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
ICN10046183 250 mg
ICN10046191 1 g
2 options

Catalog No. ICN10046183

Supplier: MP Biomedicals Inc 0210046183

Only null left

Chemical Identifiers

CAS 9004-07-3
MDL Number MFCD00130481

Specifications

Color White
Quantity 250 mg
Physical Form Powder
Chemical Name or Material α-Chymotrypsin, 3X Crystallized from Bovine Pancreas, ≥40μ/mg protein
Recommended Storage Store at +4°C