missing translation for 'onlineSavingsMsg'
Learn More

α-Chymotrypsin ≥40u/mg protein, MP Biochemicals™

Catalog No. ICN10046183
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10046183 250 mg
ICN10046191 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10046183 Supplier MP Biomedicals Inc Supplier No. 0210046183
Only null left

Chemical Identifiers

CAS 9004-07-3
MDL Number MFCD00130481

Specifications

Color White
Quantity 250 mg
Physical Form Powder
Chemical Name or Material α-Chymotrypsin, 3X Crystallized from Bovine Pancreas, ≥40μ/mg protein
Recommended Storage Store at +4°C