missing translation for 'onlineSavingsMsg'
Learn More

Chums™ Original Cotton Standard End Eyewear Retainers

Catalog No. 189992214
Encompass
Change view
Click to view available options
Color:
Assorted in Pack of 25
Black
Navy
Rainbow Tie-Dye
Red
Royal
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
189992214 Red
2980185 Assorted in Pack of 25
189992211 Black
296351180 Navy
189992213 Royal
19087035 Rainbow Tie-Dye
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 189992214 Supplier Chums™ Supplier No. 12115102
Only null left

World's first 100% cotton, fully adjustable eyewear retainer

World's first 100% cotton, fully adjustable eyewear retainer

Tough and durable, yet soft enough for everyday use.

  • Fits most standard frames
  • Logo option available
  • Made in the USA

Order Info

Available in a variety of popular colors.

Specifications

Color Red
Product Type Eyewear Retainer
For Use With (Equipment) Fits most standard frames
Material Cotton