Learn More
Eppendorf Scientific Inc CHRT RECORDER - MINI-CHRT MR
Supplier:  Eppendorf Scientific Inc P06251915
Encompass
Eppendorf TCA-3 Temperature Monitoring System is an independent temperature monitoring system for those who require complete 24/7 reliable sample protection. For those that also require alarm notification, chart recorders and autodialers can find all these features incorporated into one small pod that integrates seamlessly to the side of your freezer. This system consists of a certified RTD thermo-probe with pod measuring 6.99 cm W x 12.55 cm D x 3.24 cm H (2.75 in W x 4.94 in D x 1.28 in H) and communicates directly through any standard Ethernet connection without the need for a computer. No additional software is required to view your collected data, all that is needed is an internet browser. • Receive email, voicemail, and text alarms if the temperature exceeds desired levels for higher sample safety • Quick installation on any freezer regardless of manufacturer for flexible usage
Catalog No. 501180178
Only null left