missing translation for 'onlineSavingsMsg'
Learn More

Chromyl chloride, 99.99% (metals basis)

CAS: 14977-61-8 | Cl2CrH4O2 | 158.926 g/mol

Supplier:  Thermo Scientific Chemicals 04051709

 View more versions of this product

Catalog No. AA4051709

Only null left
Explore available promotions

Description

Description

Highly spin-polarized chromium dioxide (CrO2) thin films were deposited on 100 TiO2 substrates by chemical vapor deposition using chromyl chloride as a precursor. Chromyl chloride (Cr02C12) reacts with cyclohexane solvent at 75 OC to give a dark precipitate along with chlorocyclohexane and a small amount of cyclohexene (in 10.0 and ca. 0.3% yields based on chromium). Chromyl chloride, CrO2C12, oxidizes cyclooctane, isobutane, and toluene under mild conditions (25-60 C). The reactions give chlorinated products (chlorocyclooctane, tert- butyl chloride, and benzyl chloride) and a dark chromium-containing precipitate. Used for Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins. Chromyl chloride in carbon tetrachloride solution reacts in the cold with cyclohexene, cyclopentene, and 1-hexene to give the various isomeric chlorohydrins as the major products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins
Specifications

Specifications

117°C
-96.5°C
Liquid
(metals basis)
MFCD00010951
14,2248
Soluble in carbon tetrachloride,carbon disulfide,benzene,nitrobenzene,chloroform,and POCL3. Insoluble in water(Reacts).
O.O.[Cl-].[Cl-].[Cr+2]
158.926
154.9
Pungent, makes eyes water
1.911g/mL
Chromyl chloride
14977-61-8
10 g
Cl2CrH4O2
UN1758
chromyl chloride
HOCPFYCTFLRIAK-UHFFFAOYSA-L
chromium(2+);dichloride;dihydrate
101943071
99.99%
Moisture sensitive

Chemical Identifiers

14977-61-8
158.926
HOCPFYCTFLRIAK-UHFFFAOYSA-L
101943071
O.O.[Cl-].[Cl-].[Cr+2]
Cl2CrH4O2
MFCD00010951
chromyl chloride
chromium(2+);dichloride;dihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H340-H350-H314-H318-H317
May intensify fire
oxidizer.
May cause genetic defects.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P210-P220-P221-P260-P264b-P272-P280g-P281-P283-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P306+P360-P310-P333+P313-P353-P363-P370+P378q-P371+P380+P375-P501c

H271-H314-H317-H340-H350i

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: CHROMIUM OXYCHLORIDE

EINECSNumber : 239-056-8

RTECSNumber : GB5775000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only