missing translation for 'onlineSavingsMsg'
Learn More

Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements

Catalog No. T001010025
Change view
Click to view available options
Quantity:
25 L
Packaging:
Aluminum Jug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T001010025 25 L Aluminum Jug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T001010025 Supplier Thermo Fisher Scientific Supplier No. T001010025

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Acetonitrile is suitable for HPLC, GC, SFC, TLC, ACS, Spectrophotometry, and meets the analytical requirements of USP. | CAS: 75-05-8 | C2H3N | 41.05 g/mol

Chromplete Acetonitrile meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses

Specifications

Absorbance <0.1 at 195 nm, <0.05 at 200 nm, <0.02 at 210 nm, <0.01 at 220 nm
Density 0.775 - 0.78
Quantity 25 L
Packaging Aluminum Jug
Chemical Name or Material Acetonitrile

RUO – Research Use Only