missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chromoxane Cyanine R, pure

Catalog No. AC215150050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC215150050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC215150050 Supplier Thermo Scientific Chemicals Supplier No. 215150050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3564-18-9 | C23H15Na3O9S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 3564-18-9
Molecular Formula C23H15Na3O9S
MDL Number MFCD00012411
Synonym C.I. 43820, Mordant Blue 3

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
UV Spectrometry Complies
Beilstein 11, II, 240
Color Brown to Red
Quantity 5 g
Physical Form Powder
Formula Weight 536.39
Grade Extra Pure
Chemical Name or Material Chromoxane Cyanine R

RUO – Research Use Only