missing translation for 'onlineSavingsMsg'
Learn More

Chromotrope FB

Catalog No. AAB2232809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2232809 10 g
AAB2232818 50 g
AAB2232830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2232809 Supplier Thermo Scientific Chemicals Supplier No. B2232809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3567-69-9 | C20H12N2Na2O7S2 | 502.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3567-69-9
Molecular Formula C20H12N2Na2O7S2
Molecular Weight (g/mol) 502.42
MDL Number MFCD00003978
InChI Key OJNIRNOPCQQNRY-JZFXEGLSSA-L
Synonym Acid Red 14; C.I. 14720
PubChem CID 44135716
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=C\C(=N/NC2=C3C=CC=CC3=C(C=C2)S([O-])(=O)=O)C(=O)C2=CC=CC=C12

Specifications

Quantity 10 g
IUPAC Name disodium;4-oxo-3-[(4-sulfonatonaphthalen-1-yl)hydrazinylidene]naphthalene-1-sulfonate
Formula Weight 502.42
Chemical Name or Material Chromotrope FB
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 222-657-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only