missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chromotrope 2R, pure

CAS: 4197-07-3 | C16H10N2Na2O8S2 | 468.36 g/mol

$104.86 - $104.86

Chemical Identifiers

CAS 4197-07-3
Molecular Formula C16H10N2Na2O8S2
Molecular Weight (g/mol) 468.36
MDL Number MFCD00003955
InChI Key XTJONEUTTZZRAB-VZRGYXGKSA-L
Synonym 4,5-Dihydroxy-3-phenylazo-2,7-naphthalenedisulfonic acid, disodium salt, C.I. 16570, 4, 5-Dihydroxy-3-phenylazo-2, 7-naphthalenedisulfonic acid
PubChem CID 44135388
IUPAC Name disodium;5-hydroxy-4-oxo-3-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC192010250
SDS View Documents
Thermo Scientific Chemicals
192010250
25 g Glass bottle
Each for $104.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4197-07-3
468.36
XTJONEUTTZZRAB-VZRGYXGKSA-L
44135388
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
C16H10N2Na2O8S2
MFCD00003955
4,5-Dihydroxy-3-phenylazo-2,7-naphthalenedisulfonic acid, disodium salt, C.I. 16570, 4, 5-Dihydroxy-3-phenylazo-2, 7-naphthalenedisulfonic acid
disodium;5-hydroxy-4-oxo-3-(phenylhydrazinylidene)naphthalene-2,7-disulfonate

Specifications

4197-07-3
MFCD00003955
4,5-Dihydroxy-3-phenylazo-2,7-naphthalenedisulfonic acid, disodium salt, C.I. 16570, 4, 5-Dihydroxy-3-phenylazo-2, 7-naphthalenedisulfonic acid
Authentic
disodium;5-hydroxy-4-oxo-3-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
44135388
Extra Pure
360 min. (509nm, water)
25 g
Chromotrope 2R
C16H10N2Na2O8S2
16, 301
XTJONEUTTZZRAB-VZRGYXGKSA-L
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
468.36
468.35
Glass bottle
Brown/Red
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-085-0

RTECSNumber : QJ6418000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only