missing translation for 'onlineSavingsMsg'
Learn More

Chromosulfuric acid, Thermo Scientific Chemicals

$141.02 - $141.02

Chemical Identifiers

CAS 65272-71-1
Molecular Formula Cr2K2O7·H2O4S
Molecular Weight (g/mol) 200.237
MDL Number MFCD00010952
InChI Key IFMFTIBWWVBKOC-UHFFFAOYSA-L
Synonym absorbency linearity solution #2 24.28 mg/l inverted exclamation marka in hclo4, 25 ml
PubChem CID 131664149
IUPAC Name dipotassium;chromium;dihydroxide;dihydrate
SMILES O.O.[OH-].[OH-].[K+].[K+].[Cr]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC295510010
SDS View Documents
Thermo Scientific Chemicals
295510010
1 L Glass bottle
Each for $141.02
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

65272-71-1
200.237
IFMFTIBWWVBKOC-UHFFFAOYSA-L
131664149
O.O.[OH-].[OH-].[K+].[K+].[Cr]
Cr2K2O7·H2O4S
MFCD00010952
absorbency linearity solution #2 24.28 mg/l inverted exclamation marka in hclo4, 25 ml
dipotassium;chromium;dihydroxide;dihydrate

Specifications

Brown to Brown-Red
330.0°C
Glass bottle
1 L
Liquid
absorbency linearity solution #2 24.28 mg/l inverted exclamation marka in hclo4, 25 ml
IFMFTIBWWVBKOC-UHFFFAOYSA-L
dipotassium;chromium;dihydroxide;dihydrate
131664149
65272-71-1
1.8400g/mL
1.84
Cr2K2O7·H2O4S
MFCD00010952
Solubility in water: soluble in water (caution ! development of heat)
O.O.[OH-].[OH-].[K+].[K+].[Cr]
200.237
Chromosulfuric acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
May cause c

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only