missing translation for 'onlineSavingsMsg'
Learn More

Chromosulfuric acid, Thermo Scientific Chemicals

Catalog No. AC295510010
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295510010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC295510010 Supplier Thermo Scientific Chemicals Supplier No. 295510010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65272-71-1 | Molecular Formula: Cr2K2O7·H2O4S | Molecular Weight: 200.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 65272-71-1
Molecular Formula Cr2K2O7·H2O4S
Molecular Weight (g/mol) 200.237
MDL Number MFCD00010952
InChI Key IFMFTIBWWVBKOC-UHFFFAOYSA-L
Synonym absorbency linearity solution #2 24.28 mg/l inverted exclamation marka in hclo4, 25 ml
PubChem CID 131664149
IUPAC Name dipotassium;chromium;dihydroxide;dihydrate
SMILES O.O.[OH-].[OH-].[K+].[K+].[Cr]

Specifications

Color Brown to Brown-Red
Boiling Point 330.0°C
Density 1.8400g/mL
Packaging Glass bottle
Specific Gravity 1.84
Quantity 1 L
Physical Form Liquid
Solubility Information Solubility in water: soluble in water (caution ! development of heat)
Chemical Name or Material Chromosulfuric acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
May cause c
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only